siRNA Details
Gene Symbol: Gpr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245491.2 | XP_006245553.1 | 2 | 2920 | ||
XM_006245492.2 | XP_006245554.1 | 2 | 2538 | |||
XM_006245493.2 | XP_006245555.1 | 2 | 1725 | |||
XM_006245494.2 | XP_006245556.1 | 2 | 1067 | |||
XM_006245495.2 | XP_006245557.1 | 2 | 2656 |
