siRNA Details
Gene Symbol: St8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053914.1 | NP_446366.1 | 1, 2 | 401 | ||
GenBank mRNA | AM419015.1 | - | 1, 2 | 401 | ||
U90215.1 | - | 1, 2 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
