siRNA Details
Gene Symbol: LOC688649 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255367.2 | XP_006255429.1 | 5 | 1736 | ||
XM_006255368.2 | XP_006255430.1 | 5 | 1736 | |||
XM_006255371.2 | XP_006255433.1 | 5 | 797 | |||
XM_008772435.1 | XP_008770657.1 | 5 | 831 |
Gene Symbol: LOC102551971 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772434.1 | XP_008770656.1 | 5 | 784 |
Gene Symbol: LOC501297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772246.1 | XP_008770468.1 | 5 | 819 |
