siRNA Details
Gene Symbol: LOC501406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227820.2 | XP_006227882.1 | 4 | 739 | ||
XM_006227821.2 | XP_006227883.2 | 4 | 904 | |||
XM_008758709.1 | XP_008756931.1 | 3 | 808 |
