siRNA Details
Gene Symbol: Sh2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109313.2 | NP_001102783.2 | 2 | 368 | ||
XM_008773527.1 | XP_008771749.1 | 2 | 507 | |||
XM_008773528.1 | XP_008771750.1 | 2 | 498 | |||
GenBank mRNA | BC167036.1 | - | 2 | 368 | 7389183 |
