siRNA Details
Gene Symbol: Fancb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192003.1 | NP_001178932.1 | 7 | 2087 | ||
XM_006256884.2 | XP_006256946.1 | 8 | 2510 | |||
XM_006256885.2 | XP_006256947.1 | 9 | 2550 | |||
XM_008773184.1 | XP_008771406.1 | 8 | 2511 | |||
XM_008773185.1 | XP_008771407.1 | 10 | 2654 |
