siRNA Details
Gene Symbol: LOC501618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047970.1 | NP_001041435.1 | 1, 2 | 66 | ||
GenBank mRNA | AY539930.1 | - | 1, 2 | 66 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC501618 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047970.1 | NP_001041435.1 | 1, 2 | 66 | ||
GenBank mRNA | AY539930.1 | - | 1, 2 | 66 |