siRNA Details
Gene Symbol: Armcx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024367.1 | NP_001019538.1 | 873 | |||
XM_006257253.2 | XP_006257315.1 | 4 | 1512 | |||
GenBank mRNA | BC085780.1 | - | 873 | 7107145 | ||
FQ212524.1 | - | 835 | ||||
FQ221160.1 | - | 897 |
Gene Symbol: LOC102554790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257242.2 | XP_006257304.1 | 4 | 956 | ||
XM_006257243.2 | XP_006257305.1 | 4 | 1512 |
