siRNA Details
Gene Symbol: Fam122b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166586.1 | NP_001160058.1 | 4, 5 | 605 | ||
XM_006257643.2 | XP_006257705.1 | 4, 5 | 1145 | |||
XM_006257648.2 | XP_006257710.1 | 4, 5 | 1145 | |||
XM_008773633.1 | XP_008771855.1 | 4, 5 | 1145 | |||
XM_008773634.1 | XP_008771856.1 | 4, 5 | 1145 |
Gene Symbol: LOC100909732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752157.3 | XP_003752205.1 | 1145 | |||
XM_006257660.2 | XP_006257722.1 | 1145 | ||||
XM_006257665.2 | XP_006257727.1 | 1145 | ||||
XM_008773657.1 | XP_008771879.1 | 1145 | ||||
XM_008773658.1 | XP_008771880.1 | 1145 |
