siRNA Details
Gene Symbol: RGD1562156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768455.1 | XP_008766677.1 | 387 | |||
XM_008768456.1 | XP_008766678.1 | 2 | 469 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1562156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768455.1 | XP_008766677.1 | 387 | |||
XM_008768456.1 | XP_008766678.1 | 2 | 469 |