siRNA Details
Gene Symbol: RGD1562667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247741.2 | XP_006247803.1 | 2 | 756 | ||
XM_006247742.2 | XP_006247804.1 | 2 | 671 | |||
XM_577143.5 | XP_577143.3 | 2 | 901 |
