siRNA Details
Gene Symbol: Qrich2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247833.2 | XP_006247895.2 | 14 | 5292 | ||
XM_008768433.1 | XP_008766655.1 | 15 | 5262 | |||
XM_008768434.1 | XP_008766656.1 | 14 | 4962 | |||
XM_008768435.1 | XP_008766657.1 | 13 | 4752 | |||
XM_008768436.1 | XP_008766658.1 | 14 | 4692 | |||
XM_008768437.1 | XP_008766659.1 | 12 | 3521 |
