siRNA Details
Gene Symbol: LOC100910352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769042.1 | XP_008767264.1 | 275 |
Gene Symbol: LOC100910238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769043.1 | XP_008767265.1 | 275 |
