siRNA Details
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192006.1 | NP_001178935.1 | 2 | 225 | ||
XM_008769753.1 | XP_008767975.1 | 3 | 640 | |||
XM_008769754.1 | XP_008767976.1 | 4 | 707 | |||
XM_008769755.1 | XP_008767977.1 | 3 | 636 | |||
XM_008769756.1 | XP_008767978.1 | 3 | 638 | |||
XM_008769757.1 | XP_008767979.1 | 2 | 243 |
