siRNA Details
Gene Symbol: Cd84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192006.1 | NP_001178935.1 | 2 | 248 | ||
XM_008769753.1 | XP_008767975.1 | 3 | 663 | |||
XM_008769754.1 | XP_008767976.1 | 4 | 730 | |||
XM_008769755.1 | XP_008767977.1 | 3 | 659 | |||
XM_008769756.1 | XP_008767978.1 | 3 | 661 | |||
XM_008769757.1 | XP_008767979.1 | 2 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
