siRNA Details
Gene Symbol: RGD1562423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250509.1 | XP_006250571.1 | 733 | |||
XM_006250712.1 | XP_006250774.1 | 2 | 736 |
Gene Symbol: LOC100910292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250720.1 | XP_006250782.1 | 2 | 730 |
Gene Symbol: LOC100912233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250508.1 | XP_006250570.1 | 2 | 733 |
Gene Symbol: RGD1561699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250714.2 | XP_006250776.1 | 2 | 733 | ||
XM_008770085.1 | XP_008768307.1 | 2 | 730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
