siRNA Details
Gene Symbol: Zmat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251427.2 | XP_006251489.1 | 2 | 1444 | ||
XM_577351.5 | XP_577351.3 | 2 | 435 | |||
GenBank mRNA | FQ227485.1 | - | 2 | 187 |

More Information
Other Species Targeted

Gene Ontology Categories:
