siRNA Details
Gene Symbol: Zmat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251427.2 | XP_006251489.1 | 2 | 1385 | ||
XM_577351.5 | XP_577351.3 | 2 | 376 | |||
GenBank mRNA | FQ227485.1 | - | 2 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
