siRNA Details
Gene Symbol: Lrch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134727.1 | NP_001128199.1 | 4 | 788 | ||
XM_006252311.2 | XP_006252373.1 | 4 | 870 | |||
XM_006252312.2 | XP_006252374.1 | 4 | 864 | |||
XM_006252313.2 | XP_006252375.1 | 4 | 866 | |||
XM_006252315.2 | XP_006252377.1 | 4 | 866 | |||
XM_008770896.1 | XP_008769118.1 | 4 | 872 | |||
GenBank mRNA | BC166585.1 | - | 4 | 788 | 7444640 |

More Information
Other Species Targeted

