siRNA Details
Gene Symbol: Rwdd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034994.1 | NP_001030166.1 | 4 | 513 | ||
GenBank mRNA | BC092581.1 | - | 4 | 513 | 7369199 | |
FQ229862.1 | - | 4 | 554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
