siRNA Details
Gene Symbol: RGD1564308 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253169.2 | XP_006253231.1 | 7 | 1104 | ||
XM_006253170.2 | XP_006253232.1 | 7 | 889 | |||
XM_577523.6 | XP_577523.4 | 7 | 1120 |
Gene Symbol: LOC100911106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751598.3 | XP_003751646.2 | 2 | 313 |
