siRNA Details
Gene Symbol: Rnf122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771312.1 | XP_008769534.1 | 3, 4 | 626 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_175136, AK077856, AK171823, BC050268, BC038399, and others.
Gene Ontology Categories:
