siRNA Details
Gene Symbol: Pom121l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162931.1 | NP_001156403.1 | 1, 2 | 1598 | ||
GenBank mRNA | BC097347.1 | - | 1, 2 | 1598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
