siRNA Details
Gene Symbol: Tmem181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130939.1 | NP_001124411.1 | 5 | 426 | ||
GenBank mRNA | BC168231.1 | - | 5 | 426 | 7387910 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
