siRNA Details
Gene Symbol: LOC100912787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228762.2 | XP_006228824.1 | 5 | 1433 | ||
XM_006228763.2 | XP_006228825.1 | 5 | 1357 | |||
XM_006228764.2 | XP_006228826.1 | 5 | 1793 | |||
XM_008759233.1 | XP_008757455.1 | 5 | 1444 |
Gene Symbol: LOC103690092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759223.1 | XP_008757445.1 | 1357 |
