siRNA Details
Gene Symbol: Cdc42se2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126089.2 | NP_001119561.1 | 4 | 609 | ||
XM_006246334.1 | XP_006246396.1 | 4 | 638 | |||
XM_008767711.1 | XP_008765933.1 | 3 | 1567 | |||
GenBank mRNA | BC129107.1 | - | 4 | 609 | ||
BC169039.1 | - | 3 | 473 | 9027524 |
Gene Symbol: LOC100362023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728760.3 | XP_002728806.1 | 1 | 184 |

More Information
Other Species Targeted

Gene Ontology Categories:
