siRNA Details
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037656.1 | NP_001032745.1 | 8 | 1188 | ||
GenBank mRNA | BC107937.1 | - | 8 | 1188 | 7320504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
