siRNA Details
Gene Symbol: RGD1564915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749332.1 | XP_003749380.1 | 1 | 108 |
Gene Symbol: LOC100911153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761277.1 | XP_008759499.1 | 1 | 108 |
