siRNA Details
Gene Symbol: RGD1559714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100996.2 | NP_001094466.1 | 725 | |||
GenBank mRNA | AY902366.1 | - | 768 | |||
FJ004892.1 | - | 1265 | ||||
FJ004893.1 | - | 1579 |
Gene Symbol: LOC689173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750755.3 | XP_003750803.1 | 1265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
