siRNA Details
Gene Symbol: Mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114181.1 | NP_001107653.1 | 1 | 291 | ||
GenBank mRNA | BC158754.1 | - | 1 | 291 | 9102963 |

More Information
Other Species Targeted

Gene Ontology Categories:
