siRNA Details
Gene Symbol: Ypel4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024369.1 | NP_001019540.1 | 5 | 703 | ||
GenBank mRNA | BC083749.1 | - | 5 | 703 | 7192422 |

More Information
Other Species Targeted

