siRNA Details
Gene Symbol: Lrrc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025155.2 | NP_001020326.1 | 4 | 1406 | ||
XM_008762657.1 | XP_008760879.1 | 4 | 1694 | |||
GenBank mRNA | BC079098.1 | - | 4 | 1283 | 7124390 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028977, AK018071, BC030317.
Gene Ontology Categories:
