siRNA Details
Gene Symbol: Ankrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192012.1 | NP_001178941.1 | 1 | 144 | ||
XM_006236119.2 | XP_006236181.1 | 1 | 1377 | |||
XM_006236120.2 | XP_006236182.1 | 1 | 1377 | |||
XM_006236121.2 | XP_006236183.1 | 1 | 1377 |
