siRNA Details
Gene Symbol: Nobox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192013.1 | NP_001178942.1 | 3 | 397 | ||
XM_008762912.1 | XP_008761134.1 | 3 | 592 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nobox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192013.1 | NP_001178942.1 | 3 | 397 | ||
XM_008762912.1 | XP_008761134.1 | 3 | 592 |