siRNA Details
Gene Symbol: Kbtbd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749799.3 | XP_003749847.1 | 2 | 720 |

More Information
Other Species Targeted


UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kbtbd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749799.3 | XP_003749847.1 | 2 | 720 |