siRNA Details
Gene Symbol: Prrt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749809.3 | XP_003749857.1 | 4 | 1852 | ||
XM_006237062.2 | XP_006237124.1 | 4 | 1930 | |||
XM_006237064.2 | XP_006237126.1 | 4 | 1775 | |||
XM_008763200.1 | XP_008761422.1 | 3 | 1774 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
