siRNA Details
Gene Symbol: Clec7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173386.1 | NP_001166857.1 | 1 | 79 | ||
XM_006237100.2 | XP_006237162.1 | 1 | 202 | |||
GenBank mRNA | AB455106.1 | - | 1 | 171 | ||
AB455107.1 | - | 1 | 171 | |||
BC166774.1 | - | 1 | 79 | |||
GU357485.1 | - | 11 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
