siRNA Details
Gene Symbol: Clec7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173386.1 | NP_001166857.1 | 1 | 89 | ||
XM_006237100.2 | XP_006237162.1 | 1 | 212 | |||
GenBank mRNA | AB455106.1 | - | 1 | 181 | ||
AB455107.1 | - | 1 | 181 | |||
BC166774.1 | - | 1 | 89 | |||
GU357485.1 | - | 1 | 21 |

More Information
Other Species Targeted

Gene Ontology Categories:
