siRNA Details
Gene Symbol: Ly49si1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763315.1 | XP_008761537.1 | 7 | 1254 | ||
XM_008763316.1 | XP_008761538.1 | 7 | 1254 | |||
XM_008763318.1 | XP_008761540.1 | 6 | 1164 |
Gene Symbol: Klra17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172088.1 | NP_001165559.1 | 7 | 1003 |
Gene Symbol: LOC497796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111310.1 | NP_001104780.1 | 1159 | |||
GenBank mRNA | BC093399.1 | - | 1159 | 7389533 |
Gene Symbol: Ly49si2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763406.1 | XP_008761628.1 | 1013 |
Gene Symbol: Ly49si3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237116.2 | XP_006237178.1 | 1094 | |||
XM_008763410.1 | XP_008761632.1 | 8 | 1281 | |||
XM_008763411.1 | XP_008761633.1 | 8 | 1918 | |||
XM_008763412.1 | XP_008761634.1 | 8 | 1075 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
