siRNA Details
Gene Symbol: Angptl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025065.1 | NP_001020236.1 | 2 | 558 | ||
XM_006238440.2 | XP_006238502.1 | 2 | 745 | |||
GenBank mRNA | BC088192.1 | - | 2 | 558 | 7372069 | |
FQ210243.1 | - | 2 | 568 |
