siRNA Details
Gene Symbol: Map4k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192016.1 | NP_001178945.1 | 21, 22 | 1647 | ||
XM_006240179.2 | XP_006240241.1 | 21, 22 | 1960 | |||
XM_006240180.2 | XP_006240242.1 | 21, 22 | 1964 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_201519, BC057930, AK019468, AK032123, AK204060, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
