siRNA Details
Gene Symbol: Fam168b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271134.1 | NP_001258063.1 | 7 | 1280 | ||
XM_008766974.1 | XP_008765196.1 | 7 | 1933 | |||
XM_008766975.1 | XP_008765197.1 | 7 | 1908 | |||
XM_008766976.1 | XP_008765198.1 | 6 | 1895 | |||
XM_008766977.1 | XP_008765199.1 | 6 | 1870 | |||
XM_008766978.1 | XP_008765200.1 | 8 | 1381 | |||
XM_008766979.1 | XP_008765201.1 | 5 | 1816 |
