siRNA Details
Gene Symbol: RGD1563528 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109365.1 | NP_001102835.1 | 1 | 398 | ||
XM_006256674.2 | XP_006256736.1 | 1 | 3817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
