siRNA Details
Gene Symbol: Faim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014843.2 | NP_001014843.1 | 8 | 1298 | ||
XM_006249715.2 | XP_006249777.1 | 7 | 1364 | |||
GenBank mRNA | BC088197.1 | - | 8 | 1298 | 7375085 | |
FQ232848.1 | - | 5 | 742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
