siRNA Details
Gene Symbol: Ugt1a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039549.1 | NP_001034638.1 | 1 | 391 | ||
GenBank mRNA | AF461734.1 | - | 1 | 400 | ||
AY435132.1 | - | 1 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
