siRNA Details
Gene Symbol: Defa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033078.1 | NP_001028250.1 | 1 | 79 | ||
XM_006253369.1 | XP_006253431.1 | 2 | 177 | |||
GenBank mRNA | AY623755.1 | - | 1 | 79 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
