siRNA Details
Gene Symbol: Surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033868.1 | NP_001029040.1 | 6 | 795 | ||
GenBank mRNA | CO558944.1 | - | 1 | 116 |
Gene Symbol: LOC102553670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233722.2 | XP_006233784.1 | 5 | 2647 |
