siRNA Details
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034007.1 | NP_001029179.1 | 7 | 711 | ||
NM_001034008.1 | NP_001029180.1 | 7 | 757 | |||
XM_006252660.2 | XP_006252722.1 | 7 | 1058 | |||
GenBank mRNA | BC160817.1 | - | 7 | 736 | 7454028 | |
CV119475.1 | - | 7 | 735 |
