siRNA Details
Gene Symbol: Ncoa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034007.1 | NP_001029179.1 | 8, 9 | 1787 | ||
NM_001034008.1 | NP_001029180.1 | 8, 9 | 1833 | |||
XM_006252660.2 | XP_006252722.1 | 8, 9 | 2134 | |||
GenBank mRNA | BC160817.1 | - | 8, 9 | 1812 | 7454028 |
