siRNA Details
Gene Symbol: Med24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034079.1 | NP_001029251.1 | 19 | 1910 | ||
XM_006247415.2 | XP_006247477.1 | 20 | 2007 | |||
XM_006247416.2 | XP_006247478.1 | 20 | 2053 | |||
GenBank mRNA | BC097461.1 | - | 19 | 1910 | 7457533 | |
FQ210692.1 | - | 19 | 1933 |

More Information
Other Species Targeted

Gene Ontology Categories:
